missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Fluorophenyl)-1-phenyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H64626.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15461909

  • £59.78 / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

3-(4-Fluorophenyl)-1-phenyl-2-thiohydantoin
286.32
MFCD27665295
3-(4-Fluorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone

Specifications

3-(4-Fluorophenyl)-1-phenyl-2-thiohydantoin
Solid
286.32
3-(4-Fluorophenyl)-1-phenyl-2-thioxo-4-imidazolidinone
1g
95%
MFCD27665295
Videos
SDS