missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Fluorophenyl)-1-methyl-2-thiohydantoin, 95%

Brand:  Alfa Aesar™ H63504.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15410239

  • £373.12 / 5g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

3-(4-Fluorophenyl)-1-methyl-2-thiohydantoin
224.25
MFCD06676158
3-(4-Fluorophenyl)-1-methyl-2-thioxo-4-imidazolidinone

Specifications

3-(4-Fluorophenyl)-1-methyl-2-thiohydantoin
Solid
224.25
3-(4-Fluorophenyl)-1-methyl-2-thioxo-4-imidazolidinone
5g
95%
MFCD06676158
Videos
SDS