missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorophenyl)-1-isopropylindole, 98%, Thermo Scientific™

Catalog Number p-7046793
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
93957-49-4
C17H16FN
253.32
MFCD01075733
ZDZJOIIBECYKAJ-UHFFFAOYSA-N
3-4-fluorophenyl-1-isopropyl-1h-indole, 1-isopropyl-3-4-fluorophenyl indole, 3-4-fluorophenyl-1-isopropylindole, 3-4-fluorophenyl-1-1-methylethyl-1h-indole, 1-isopropyl-3-4-flrorophenyl-indole, 1h-indole, 3-4-fluorophenyl-1-1-methylethyl, 4-fluorophenyl-1-isopropyl-3-indole, 3-4-fluorophenyl-1-propan-2-ylindole, 3-4-fluoro-phenyl-1-isopropyl-1h-indole, 3-4-fluorophenyl-1-methylethyl indole
736229
3-(4-fluorophenyl)-1-propan-2-ylindole
CC(C)N1C=C(C2=CC=CC=C21)C3=CC=C(C=C3)F
This item is not returnable. View return policy
93957-49-4
C17H16FN
253.32
MFCD01075733
ZDZJOIIBECYKAJ-UHFFFAOYSA-N
3-4-fluorophenyl-1-isopropyl-1h-indole, 1-isopropyl-3-4-fluorophenyl indole, 3-4-fluorophenyl-1-isopropylindole, 3-4-fluorophenyl-1-1-methylethyl-1h-indole, 1-isopropyl-3-4-flrorophenyl-indole, 1h-indole, 3-4-fluorophenyl-1-1-methylethyl, 4-fluorophenyl-1-isopropyl-3-indole, 3-4-fluorophenyl-1-propan-2-ylindole, 3-4-fluoro-phenyl-1-isopropyl-1h-indole, 3-4-fluorophenyl-1-methylethyl indole
736229
3-(4-fluorophenyl)-1-propan-2-ylindole
CC(C)N1C=C(C2=CC=CC=C21)C3=CC=C(C=C3)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 95°C to 98°C
Quantity 5 g
Beilstein 4457093
Formula Weight 253.32
Percent Purity 98%
Chemical Name or Material 3-(4-Fluorophenyl)-1-isopropylindole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'