missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenylboronic acid, 97%, ACROS Organics™

Brand:  Acros Organics 359280050

Code : 29

Additional Details : CAS Number : 55499-43-9 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10456432

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3, 4-Dimethylphenylboronic acid
96.0
97%
C8H11BO2
MFCD01009694
55499-43-9
100.0
97%
149.98

Specifications

3, 4-Dimethylphenylboronic acid
5g
96.0
100.0
Powder
97%
149.98
White to slightly beige
55499-43-9
Authentic
Glass bottle
97%
C8H11BO2
MFCD01009694
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,4-Dimethylphenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS