missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4-Dimethylphenol, 98+%

Brand:  Alfa Aesar™ A10599.36

Code : NEW

Additional Details : CAS Number : 95-65-8 Weight : 0.50000kg Transport : UN number : 2261 Chem class : 1 Pack group : II

Quantity 500g

 View more versions of this product

Product Code. 11493274

  • £78.10 / 500g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Description

Description

Applications
3,4-Dimethylphenol, is used as an anti-Infective agents. It has wide applications in the analytical techniques such as Gas Chromatography, Reversed Phase Chromatography, HPLC and UHPLC.

Solubility
Slightly soluble in water. Less than 1 mg/mL at 68° F.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Light sensitive.Hygroscopic. Incompatible with bases, acid chlorides, acid anhydrides, and oxidizing agents.
Specifications

Chemical Identifiers

3,4-Dimethylphenol
C8H10O
MFCD00002304
1099267
3,4-Xylenol
95-65-8
122.17
UN2261
14,10082

Specifications

3,4-Dimethylphenol
65° to 68°C
95-65-8
C8H10O
MFCD00002304
1099267
3,4-Xylenol
227°C
500g
≥98%
122.17
UN2261
14,10082
Videos
SDS