missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number 10576441
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5973-71-7
C9H10O
134.18
MFCD00016612
POQJHLBMLVTHAU-UHFFFAOYSA-N
benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehyde
22278
3,4-dimethylbenzaldehyde
CC1=C(C=C(C=C1)C=O)C
This item is not returnable. View return policy
5973-71-7
C9H10O
134.18
MFCD00016612
POQJHLBMLVTHAU-UHFFFAOYSA-N
benzaldehyde, 3,4-dimethyl, unii-16g1j12arb, 4-formyl-o-xylene, o-xylene-4-carboxaldehyde, 3,4-dimethyl-benzaldehyde, 3 pound not4-dimethylbenzaldehyde, pubchem2535, 3,4 dimethyl benzaldehyde, 3,4-di-methylbenzaldehyde, 3,4-dimethyl benzaldehyde
22278
3,4-dimethylbenzaldehyde
CC1=C(C=C(C=C1)C=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0120g/mL
Boiling Point 226°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C6H3CHO
Refractive Index 1.5490 to 1.551
Quantity 5 g
Beilstein 07,IV,707
Specific Gravity 1.012
Formula Weight 134.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dimethylbenzaldehyde, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'