missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethylaniline, 99.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 49°C to 51°C |
CAS Min % | 99.4 |
Color | Beige to Brown |
CAS Max % | 100.0 |
Assay Percent Range | 99.4% min. (GC) |
Linear Formula | (CH3)2C6H3NH2 |
Beilstein | 12, 1103 |
Merck Index | 15, 10282 |
Solubility Information | Solubility in water: <1g/L (24°C). Other solubilities: slightly soluble in chloroform,soluble in alcohol (>100g/L),diethyl ether,,petroleum ether,very soluble in ligroin |
Formula Weight | 121.18 |
Show More |
Safety and Handling
Product Identifier
- 3,4-Dimethylaniline
- Danger
- Acute toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H311-Toxic in contact with skin.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'