missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethylacetophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-7081972
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
3637-01-2
C10H12O
148.21
MFCD00008743
WPRAXAOJIODQJR-UHFFFAOYSA-N
This item is not returnable. View return policy
3637-01-2
C10H12O
148.21
MFCD00008743
WPRAXAOJIODQJR-UHFFFAOYSA-N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.002
Boiling Point 242°C to 243°C
Flash Point 102°C (215°F)
Refractive Index 1.538
Quantity 5 g
Beilstein 907053
Synonym 3',4'-dimethylacetophenone, 1-3,4-dimethylphenyl ethanone, 3,4-dimethylacetophenone, ethanone, 1-3,4-dimethylphenyl, 1-3,4-dimethylphenyl ethan-1-one, unii-pc1l145t5v, 1-3,4-dimethyl-phenyl-ethanone, 1-acetyl-3,4-dimethylbenzene, 3.4-dimethylacetophenon, acmc-1cmy2
SMILES CC1=C(C=C(C=C1)C(=O)C)C
IUPAC Name 1-(3,4-dimethylphenyl)ethanone
PubChem CID 77193
Formula Weight 148.21
Percent Purity 98%
Chemical Name or Material 3',4'-Dimethylacetophenone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'