missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxyphenylacetonitrile, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
It is employed as an intermediate in the preparation of the muscle relaxant papverin. 3,4-Dimethoxyphenyl)acetonitrile has been used in the synthesis of (Z)-3-(benzo[b]thiophen-2-yl)-2-(3,4-dimethoxyphenyl)acrylonitrile1. It has also been used in the preparation of modified diterpene (±) nimbidiol which relies on a sulfenium ion promoted polyene cyclization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as an intermediate in the preparation of the muscle relaxant papverin. 3,4-Dimethoxyphenyl)acetonitrile has been used in the synthesis of (Z)-3-(benzo[b]thiophen-2-yl)-2-(3,4-dimethoxyphenyl)acrylonitrile1. It has also been used in the preparation of modified diterpene (±) nimbidiol which relies on a sulfenium ion promoted polyene cyclization.
Solubility
Insoluble in water. Soluble in methanol.
Notes
Store in cool place. Keep away from oxidizing agents.
It is employed as an intermediate in the preparation of the muscle relaxant papverin. 3,4-Dimethoxyphenyl)acetonitrile has been used in the synthesis of (Z)-3-(benzo[b]thiophen-2-yl)-2-(3,4-dimethoxyphenyl)acrylonitrile1. It has also been used in the preparation of modified diterpene (±) nimbidiol which relies on a sulfenium ion promoted polyene cyclization.
Solubility
Insoluble in water. Soluble in methanol.
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
Melting Point | 62°C to 65°C |
Boiling Point | 177°C to 178°C (10 mmHg) |
Odor | Characteristic |
Quantity | 25 g |
Beilstein | 1956100 |
Solubility Information | Insoluble in water. Soluble in methanol. |
Formula Weight | 177.2 |
Percent Purity | 98% |
Chemical Name or Material | 3,4-Dimethoxyphenylacetonitrile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'