missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dimethoxyphenyl isothiocyanate, 97%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 10g
Specifications
CAS | 33904-04-0 |
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Molecular Formula | C9H9NO2S |
MDL Number | MFCD00041075 |
Quantity | 10g |
Synonym | 3,4-dimethoxyphenyl isothiocyanate, 3,4-dimethoxyphenylisothiocyanate, 4-isothiocyanato-1,2-dimethoxy-benzene, isothiocyanic acid 3,4-dimethoxyphenyl ester, 3,4-dimethoxybenzenisothiocyanate, acmc-1aeuy, 3,4 dimethoxyphenylisothiocyanate, lhpzzvzpozpddb-uhfffaoysa, #, 3,4-dimethoxy phenyl isothiocyanate |
InChI Key | LHPZZVZPOZPDDB-UHFFFAOYSA-N |
SMILES | COC1=C(C=C(C=C1)N=C=S)OC |
Show More |
Safety and Handling
Product Identifier
- 3,4-Dimethoxyphenyl isothiocyanate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'