missing translation for 'onlineSavingsMsg'
Learn More

(3,4-Dimethoxyphenyl)acetic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-114030
13.60 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
93-40-3
C10H12O4
196.2
MFCD00004335
WUAXWQRULBZETB-UHFFFAOYSA-N
homoveratric acid, 3,4-dimethoxyphenylacetic acid, 3,4-dimethoxyphenyl acetic acid, 2-3,4-dimethoxyphenyl acetic acid, benzeneacetic acid, 3,4-dimethoxy, 3,4-dimethoxybenzeneacetic acid, 3,4-dimethoxy-phenyl-acetic acid, acetic acid, 3,4-dimethoxyphenyl
7139
CHEBI:86655
2-(3,4-dimethoxyphenyl)acetic acid
COC1=C(C=C(C=C1)CC(=O)O)OC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
93-40-3
C10H12O4
196.2
MFCD00004335
WUAXWQRULBZETB-UHFFFAOYSA-N
homoveratric acid, 3,4-dimethoxyphenylacetic acid, 3,4-dimethoxyphenyl acetic acid, 2-3,4-dimethoxyphenyl acetic acid, benzeneacetic acid, 3,4-dimethoxy, 3,4-dimethoxybenzeneacetic acid, 3,4-dimethoxy-phenyl-acetic acid, acetic acid, 3,4-dimethoxyphenyl
7139
CHEBI:86655
2-(3,4-dimethoxyphenyl)acetic acid
COC1=C(C=C(C=C1)CC(=O)O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 96°C to 99°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula (CH3O)2C6H3CH2CO2H
Quantity 25 g
Beilstein 10, 409
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 196.2
Percent Purity 99%
Physical Form Powder
Chemical Name or Material (3, 4-Dimethoxyphenyl)acetic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • (3,4-Dimethoxyphenyl)acetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'