3,4-Dimethoxyphenethylamine, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 115621000

UNSPSC: 12350000

Code: X2

Additional Details:
Additional Details: CAS Number: 120-20-7 Weight: 0.15000kg


Packaging Glass bottle
Quantity 100g

Product Code. 10204290

Quantity Price
1 £ 47.2 / 100g
Estimated Shipment
23-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3, 4-Dimethoxyphenethylamine
CAS 120-20-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H15NO2
Linear Formula (CH3O)2C6H3CH2CH2NH2
Formula Weight 181.23
MDL Number MFCD00008188

Specification

Formula Weight 181.23
Color Clear light yellow to orange or slightly brownish
Physical Form Viscous Liquid
Assay ≥97.5%
Quantity 100g
Beilstein 13, 800
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula (CH3O)2C6H3CH2CH2NH2
Infrared Spectrum Authentic
CAS 120-20-7
MDL Number MFCD00008188
Refractive Index 1.5450 to 1.5470 (20°C, 589nm)
Assay Percent Range 98%
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
CAS Max % 100.0
Density 1.0700g/mL
Molecular Formula C10H15NO2
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material 3, 4-Dimethoxyphenethylamine
Synonym 2-(3, 4-Dimethoxyphenyl)ethylamine
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.