missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxycinnamic acid, 99%, predominantly trans isomer, Thermo Scientific Chemicals

Catalog Number p-114557
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2316-26-9
C11H12O4
208.21
MFCD00004387
HJBWJAPEBGSQPR-GQCTYLIASA-N
3,4-dimethoxycinnamic acid, dimethylcaffeic acid, caffeic acid dimethyl ether, 2e-3-3,4-dimethoxyphenyl prop-2-enoic acid, dimethyl caffeic acid, e-3-3,4-dimethoxyphenyl acrylic acid, e-3,4-dimethoxycinnamic acid, unii-bvz841pvjl, 2e-3-3,4-dimethoxyphenyl acrylic acid, 3-3,4-dimethoxyphenyl acrylic acid
717531
CHEBI:86549
(E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
COC1=C(C=C(C=C1)C=CC(=O)O)OC
This item is not returnable. View return policy
2316-26-9
C11H12O4
208.21
MFCD00004387
HJBWJAPEBGSQPR-GQCTYLIASA-N
3,4-dimethoxycinnamic acid, dimethylcaffeic acid, caffeic acid dimethyl ether, 2e-3-3,4-dimethoxyphenyl prop-2-enoic acid, dimethyl caffeic acid, e-3-3,4-dimethoxyphenyl acrylic acid, e-3,4-dimethoxycinnamic acid, unii-bvz841pvjl, 2e-3-3,4-dimethoxyphenyl acrylic acid, 3-3,4-dimethoxyphenyl acrylic acid
717531
CHEBI:86549
(E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
COC1=C(C=C(C=C1)C=CC(=O)O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 181°C to 184°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH=CHCO2H
Quantity 5 g
Beilstein 10, 437
Formula Weight 208.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3, 4-Dimethoxycinnamic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxycinnamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'