missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzyl alcohol, 96%, Thermo Scientific Chemicals

Catalog Number 11934731
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
93-03-8
C9H12O3
168.19
OEGPRYNGFWGMMV-UHFFFAOYSA-N
3,4-dimethoxybenzyl alcohol, veratryl alcohol, 3,4-dimethoxyphenyl methanol, benzenemethanol, 3,4-dimethoxy, 3,4-dimethoxyphenylmethyl alcohol, veratrole alcohol, 3,4-dimethoxybenzenemethanol, unii-mb4t4a711h, 3,4-dimethoxyphenyl-methanol
7118
CHEBI:62150
(3,4-dimethoxyphenyl)methanol
COC1=C(C=C(C=C1)CO)OC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
93-03-8
C9H12O3
168.19
OEGPRYNGFWGMMV-UHFFFAOYSA-N
3,4-dimethoxybenzyl alcohol, veratryl alcohol, 3,4-dimethoxyphenyl methanol, benzenemethanol, 3,4-dimethoxy, 3,4-dimethoxyphenylmethyl alcohol, veratrole alcohol, 3,4-dimethoxybenzenemethanol, unii-mb4t4a711h, 3,4-dimethoxyphenyl-methanol
7118
CHEBI:62150
(3,4-dimethoxyphenyl)methanol
COC1=C(C=C(C=C1)CO)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 22.0°C
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 296.0°C to 297.0°C (732.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH2OH
Refractive Index 1.5510 to 1.5540
Quantity 500 g
Beilstein 06,1113
Specific Gravity 1.15
Solubility Information Solubility in water: miscible
Formula Weight 168.19
Percent Purity 96%
Physical Form Oily Liquid
Chemical Name or Material 3, 4-Dimethoxybenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'