missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzonitrile, 98+%, Thermo Scientific Chemicals

Catalog Number p-7020532
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
2024-83-1
C9H9NO2
163.18
MFCD00001802
OSEQIDSFSBWXRE-UHFFFAOYSA-N
veratronitrile, 4-cyanoveratrole, 3,4-dimethoxy-benzonitrile, 4-cyano-1,2-dimethoxybenzene, 1-cyano-3,4-dimethoxylbenzene, 3,4-dimethoxybenzoic acid nitrile, 3,4-dimethoxyphenyl cyanide, benzonitrile,4-dimethoxy, 3,4-dimethoxybenzenecarbonitrile, 3-cyanoveratrole
74842
3,4-dimethoxybenzonitrile
COC1=CC=C(C=C1OC)C#N
This item is not returnable. View return policy
2024-83-1
C9H9NO2
163.18
MFCD00001802
OSEQIDSFSBWXRE-UHFFFAOYSA-N
veratronitrile, 4-cyanoveratrole, 3,4-dimethoxy-benzonitrile, 4-cyano-1,2-dimethoxybenzene, 1-cyano-3,4-dimethoxylbenzene, 3,4-dimethoxybenzoic acid nitrile, 3,4-dimethoxyphenyl cyanide, benzonitrile,4-dimethoxy, 3,4-dimethoxybenzenecarbonitrile, 3-cyanoveratrole
74842
3,4-dimethoxybenzonitrile
COC1=CC=C(C=C1OC)C#N

3,4-Dimethoxybenzonitrile is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxybenzonitrile is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 65°C to 69°C
Quantity 25 g
UN Number UN3439
Beilstein 1451651
Formula Weight 163.18
Percent Purity ≥98%
Chemical Name or Material 3,4-Dimethoxybenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'