missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7027465
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
122775-35-3
C8H11BO4
181.98
MFCD01074574
RCVDPBFUMYUKPB-UHFFFAOYSA-N
3,4-dimethoxyphenyl boronic acid, 3,4-dimethoxybenzeneboronic acid, 3,4-dimethoxyphenyl boranediol, 3,4-dimethoxybenzene boronic acid, boronic acid, 3,4-dimethoxyphenyl, 3,4-dimethoxyphenylboronicacid, 3, 4-dimethoxyphenylboronic acid, pubchem1826, acmc-209amm, ksc174q5t
2734702
(3,4-dimethoxyphenyl)boronic acid
COC1=CC=C(C=C1OC)B(O)O
This item is not returnable. View return policy
122775-35-3
C8H11BO4
181.98
MFCD01074574
RCVDPBFUMYUKPB-UHFFFAOYSA-N
3,4-dimethoxyphenyl boronic acid, 3,4-dimethoxybenzeneboronic acid, 3,4-dimethoxyphenyl boranediol, 3,4-dimethoxybenzene boronic acid, boronic acid, 3,4-dimethoxyphenyl, 3,4-dimethoxyphenylboronicacid, 3, 4-dimethoxyphenylboronic acid, pubchem1826, acmc-209amm, ksc174q5t
2734702
(3,4-dimethoxyphenyl)boronic acid
COC1=CC=C(C=C1OC)B(O)O

3,4-Dimethoxybenzeneboronic acid is used in organic light-emitting diode. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxybenzeneboronic acid is used in organic light-emitting diode. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in water (25 g/L).

Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼240°C
Quantity 1 g
Beilstein 5334877
Solubility Information Soluble in water (25g/L).
Formula Weight 181.98
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxybenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'