missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzaldehyde, 99+%, Thermo Scientific Chemicals

Catalog Number p-114517
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
120-14-9
C9H10O3
166.18
MFCD00003363
WJUFSDZVCOTFON-UHFFFAOYSA-N
veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin
8419
CHEBI:17098
3,4-dimethoxybenzaldehyde
COC1=CC=C(C=O)C=C1OC
This item is not returnable. View return policy
120-14-9
C9H10O3
166.18
MFCD00003363
WJUFSDZVCOTFON-UHFFFAOYSA-N
veratraldehyde, veratric aldehyde, methylvanillin, vanillin methyl ether, veratral, veratryl aldehyde, benzaldehyde, 3,4-dimethoxy, veratrum aldehyde, p-veratric aldehyde, 4-o-methylvanillin
8419
CHEBI:17098
3,4-dimethoxybenzaldehyde
COC1=CC=C(C=O)C=C1OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point 42°C to 45°C
Color White to Yellow
Boiling Point 281°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3O)2C6H3CHO
Quantity 5 g
Beilstein 08, 255
Merck Index 15, 10145
Solubility Information Solubility in water: soluble in hot water. Other solubilities: freely soluble in alcohol and ether
Formula Weight 166.18
Percent Purity 99+%
Physical Form Lumps or Fused Solid
Chemical Name or Material 3, 4-Dimethoxybenzaldehyde, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'