missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxy-N-methylbenzylamine, 97%, Thermo Scientific™

Catalog Number 11945311
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
63-64-9
C10H15NO2
181.24
XQODFBIAQVJQHF-UHFFFAOYSA-N
3,4-dimethoxybenzyl methylamine, n-methyl-3,4-dimethoxybenzylamine, 1-3,4-dimethoxyphenyl-n-methylmethanamine, 3,4-dimethoxyphenyl methyl methyl amine, 3,4-dimethoxy-n-methylbenzylamine, 3,4-dimethoxy-benzyl-methyl-amine, benzenemethanamine, 3,4-dimethoxy-n-methyl, 1-3,4-dimethoxyphenyl-n-methyl-methanamine, 3,4-dimethoxyphenyl methyl methylamine, 3,4-dimethoxybenzyl-n-methylamine
592161
1-(3,4-dimethoxyphenyl)-N-methylmethanamine
CNCC1=CC(=C(C=C1)OC)OC
This item is not returnable. View return policy
63-64-9
C10H15NO2
181.24
XQODFBIAQVJQHF-UHFFFAOYSA-N
3,4-dimethoxybenzyl methylamine, n-methyl-3,4-dimethoxybenzylamine, 1-3,4-dimethoxyphenyl-n-methylmethanamine, 3,4-dimethoxyphenyl methyl methyl amine, 3,4-dimethoxy-n-methylbenzylamine, 3,4-dimethoxy-benzyl-methyl-amine, benzenemethanamine, 3,4-dimethoxy-n-methyl, 1-3,4-dimethoxyphenyl-n-methyl-methanamine, 3,4-dimethoxyphenyl methyl methylamine, 3,4-dimethoxybenzyl-n-methylamine
592161
1-(3,4-dimethoxyphenyl)-N-methylmethanamine
CNCC1=CC(=C(C=C1)OC)OC
CAS Min % 96.0
CAS Max % 100.0
Color Brown to Colorless
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 181.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dimethoxy-N-methylbenzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxy-N-methylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'