missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzylamine hydrobromide, 98%, Thermo Scientific Chemicals

Catalog Number 10561861
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
16290-26-9
C7H10BrNO2
220.07
MFCD00012859
BVFZTXFCZAXSHN-UHFFFAOYSA-N
3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, dhba hydrobromide, acmc-1afa4, 1, 4-aminomethyl-, hydrobromide, 4-aminomethyl catechol hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide
13343562
4-(aminomethyl)benzene-1,2-diol;hydrobromide
[H+].[Br-].NCC1=CC=C(O)C(O)=C1
This item is not returnable. View return policy
16290-26-9
C7H10BrNO2
220.07
MFCD00012859
BVFZTXFCZAXSHN-UHFFFAOYSA-N
3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, dhba hydrobromide, acmc-1afa4, 1, 4-aminomethyl-, hydrobromide, 4-aminomethyl catechol hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide
13343562
4-(aminomethyl)benzene-1,2-diol;hydrobromide
[H+].[Br-].NCC1=CC=C(O)C(O)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point >180.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (HO)2C6H3CH2NH2·HBr
Quantity 250 mg
Beilstein 13, 796
Formula Weight 220.07
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dihydroxybenzylamine hydrobromide, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'