missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 3,4-Dihydroxy-DL-phenylalanine, 98%

Catalog Number p-7051561
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
63-84-3
C9H11NO4
197.19
MFCD00063060
WTDRDQBEARUVNC-UHFFFAOYNA-N
dl-dopa, 3,4-dihydroxy-dl-phenylalanine, 2-amino-3-3,4-dihydroxyphenyl propanoic acid, dl-dioxyphenylalanine, dl-dihydroxyphenylalanine, 3-3,4-dihydroxyphenyl-dl-alanine, 3-hydroxytyrosine, 3-hydroxy-dl-tyrosine, dopa, dl-3,4-dopa
836
CHEBI:49168
2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
NC(CC1=CC=C(O)C(O)=C1)C(O)=O
This item is not returnable. View return policy
63-84-3
C9H11NO4
197.19
MFCD00063060
WTDRDQBEARUVNC-UHFFFAOYNA-N
dl-dopa, 3,4-dihydroxy-dl-phenylalanine, 2-amino-3-3,4-dihydroxyphenyl propanoic acid, dl-dioxyphenylalanine, dl-dihydroxyphenylalanine, 3-3,4-dihydroxyphenyl-dl-alanine, 3-hydroxytyrosine, 3-hydroxy-dl-tyrosine, dopa, dl-3,4-dopa
836
CHEBI:49168
2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
NC(CC1=CC=C(O)C(O)=C1)C(O)=O

3,4-Dihydroxy-DL-phenylalanine is used as a dopamine precursor. It is also useful in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 290°C (decomposition)
Merck Index 14,3420
Solubility Information Soluble in water,0.5M dilute hydrochloric acid and formic acid. Insoluble in ethanol,benzene,chloroform and ethyl acetate.
Formula Weight 197.19
Percent Purity 98%
Quantity 5 g
Odor Odorless
Sensitivity Air Sensitive
Physical Form Powder
Chemical Name or Material 3,4-Dihydroxy-DL-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'