missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydro-2H-1,5-benzodioxepin-7-ylboronic acid, 90+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 193.99 |
Chemical Name or Material | 3, 4-Dihydro-2H-1, 5-benzodioxepin-7-ylboronic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'