missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-6-ylmethylamine hydrochloride, 97%, Thermo Scientific™

Catalog Number p-2698894
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
499770-91-1
C10H13NO2
179.22
MFCD04972615
ZXJOKQNPRXXPJT-UHFFFAOYSA-N
3,4-dihydro-2h-1,5-benzodioxepin-6-ylmethylamine, 3,4-dihydro-2h-benzo b 1,4 dioxepin-6-yl methanamine, 1-3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, 2h-1,5-benzodioxepin-6-methanamine,3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, vxu
2794995
NCC1=C2OCCCOC2=CC=C1
This item is not returnable. View return policy
499770-91-1
C10H13NO2
179.22
MFCD04972615
ZXJOKQNPRXXPJT-UHFFFAOYSA-N
3,4-dihydro-2h-1,5-benzodioxepin-6-ylmethylamine, 3,4-dihydro-2h-benzo b 1,4 dioxepin-6-yl methanamine, 1-3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, 2h-1,5-benzodioxepin-6-methanamine,3,4-dihydro, 3,4-dihydro-2h-1,5-benzodioxepin-6-yl methanamine, vxu
2794995
NCC1=C2OCCCOC2=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 1-(3,4-dihydro-2H-1,5-benzodioxepin-6-yl)methanamine
Formula Weight 215.68
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepin-6-ylmethylamine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'