missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester, Thermo Scientific Chemicals

Catalog Number p-7054242
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1155264-46-2
C14H20BNO3
261.13
MFCD18073255
HFUHUNYUUCDCAU-UHFFFAOYSA-N
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, acmc-2099qf, 3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazine, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine
54759084
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
CC1(C)OB(OC1(C)C)C1=CC2=C(OCCN2)C=C1
This item is not returnable. View return policy
1155264-46-2
C14H20BNO3
261.13
MFCD18073255
HFUHUNYUUCDCAU-UHFFFAOYSA-N
6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, acmc-2099qf, 3,4-dihydro-2h-benzo b 1,4 oxazin-6-yl boronic acid pinacol ester, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-1,4-benzoxazine, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 6-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine
54759084
6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
CC1(C)OB(OC1(C)C)C1=CC2=C(OCCN2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 261.12
Physical Form Solid
Chemical Name or Material 3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'