missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,4-benzoxazine-2-carbonitrile, 97+%, Thermo Scientific™

Catalog Number p-4510355
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
86267-86-9
C9H8N2O
160.176
YSTANLOUKDVPGJ-UHFFFAOYSA-N
3,4-dihydro-2h-benzo b 1,4 oxazine-2-carbonitrile, 2h,3h,4h-benzo e 1,4-oxazaperhydroine-2-carbonitrile, 2h-1,4-benzoxazine-2-carbonitrile,3,4-dihydro
2795504
3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile
C1C(OC2=CC=CC=C2N1)C#N
This item is not returnable. View return policy
86267-86-9
C9H8N2O
160.176
YSTANLOUKDVPGJ-UHFFFAOYSA-N
3,4-dihydro-2h-benzo b 1,4 oxazine-2-carbonitrile, 2h,3h,4h-benzo e 1,4-oxazaperhydroine-2-carbonitrile, 2h-1,4-benzoxazine-2-carbonitrile,3,4-dihydro
2795504
3,4-dihydro-2H-1,4-benzoxazine-2-carbonitrile
C1C(OC2=CC=CC=C2N1)C#N
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 160.18
Percent Purity 97%
Chemical Name or Material 3, 4-Dihydro-2H-1, 4-benzoxazine-2-carbonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'