missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2(1H)-quinolinone, 98%, Thermo Scientific Chemicals

Catalog Number p-3759042
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
553-03-7
C9H9NO
147.18
MFCD00016722
TZOYXRMEFDYWDQ-UHFFFAOYSA-N
3,4-dihydro-2 1h-quinolinone, 3,4-dihydroquinolin-2 1h-one, hydrocarbostyril, 1,2,3,4-tetrahydroquinolin-2-one, 3,4-dihydrocarbostyril, 2-oxo-1,2,3,4-tetrahydroquinoline, dihydro-quinolinone, 3,4-dihydro-2-quinolinol, o-aminohydrocinnamic acid lactam, unii-2ckg6tx32f
64796
3,4-dihydro-1H-quinolin-2-one
O=C1CCC2=CC=CC=C2N1
This item is not returnable. View return policy
553-03-7
C9H9NO
147.18
MFCD00016722
TZOYXRMEFDYWDQ-UHFFFAOYSA-N
3,4-dihydro-2 1h-quinolinone, 3,4-dihydroquinolin-2 1h-one, hydrocarbostyril, 1,2,3,4-tetrahydroquinolin-2-one, 3,4-dihydrocarbostyril, 2-oxo-1,2,3,4-tetrahydroquinoline, dihydro-quinolinone, 3,4-dihydro-2-quinolinol, o-aminohydrocinnamic acid lactam, unii-2ckg6tx32f
64796
3,4-dihydro-1H-quinolin-2-one
O=C1CCC2=CC=CC=C2N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 165.0°C to 167.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 4817
Formula Weight 147.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dihydro-2(1H)-quinolinone
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dihydro-2(1H)-quinolinone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'