missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10531384
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
168267-41-2
C6H5BF2O2
157.91
MFCD00807405
RMGYQBHKEWWTOY-UHFFFAOYSA-N
3,4-difluorophenyl boronic acid, 3,4-difluorobenzeneboronic acid, 3,4-difluorophenyl boranediol, 3,4-fluorobenzene boronic acid, 3,4-difluorophenylbornic acid, 3,4-difluoro phenylboric acid, 3,4-difluorobenzenebornoic acid, 3,4-difluoro phenylboronic acid, boronic acid, 3,4-difluorophenyl
2734337
(3,4-difluorophenyl)boronic acid
OB(O)C1=CC=C(F)C(F)=C1
This item is not returnable. View return policy
168267-41-2
C6H5BF2O2
157.91
MFCD00807405
RMGYQBHKEWWTOY-UHFFFAOYSA-N
3,4-difluorophenyl boronic acid, 3,4-difluorobenzeneboronic acid, 3,4-difluorophenyl boranediol, 3,4-fluorobenzene boronic acid, 3,4-difluorophenylbornic acid, 3,4-difluoro phenylboric acid, 3,4-difluorobenzenebornoic acid, 3,4-difluoro phenylboronic acid, boronic acid, 3,4-difluorophenyl
2734337
(3,4-difluorophenyl)boronic acid
OB(O)C1=CC=C(F)C(F)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 289°C to 290°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 157.91
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3, 4-Difluorophenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Difluorophenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'