missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzaldehyde, 97%, Maybridge

Brand:  Maybridge SB01773FA

Code : 31

Additional Details : CAS Number : 34036-07-2 Weight : 0.15000kg

Quantity 100g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10198744

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

3, 4-Difluorobenzaldehyde
97%
F2C6H3CHO
97%
C7H4F2O
142.1

Specifications

3, 4-Difluorobenzaldehyde
100g
Amber Glass Bottle
97%
F2C6H3CHO
1.2900g/mL
34036-07-2
97%
C7H4F2O
142.1
Videos
SDS