missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4-Difluoro-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61736.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496746

  • £56.29 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

3,4-Difluoro-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide
337.27
MFCD05116135

Specifications

3,4-Difluoro-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide
1g
97%
MFCD05116135
Orange
Solid
337.27
Videos
SDS