missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4-Difluoro-N-(3-fluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61946.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486876

  • £73.56 / 1g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

3,4-Difluoro-N-(3-fluorophenyl)benzenesulfonamide
287.26
MFCD06022643

Specifications

3,4-Difluoro-N-(3-fluorophenyl)benzenesulfonamide
Solid
287.26
1g
97%
MFCD06022643
Videos
SDS