missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4-Difluoro-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61211.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15426356

  • £86.92 / 1g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

3,4-Difluoro-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
337.27
MFCD05116078

Specifications

3,4-Difluoro-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
1g
97%
MFCD05116078
Pale orange
Solid
337.27
Videos
SDS