missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-5-methylbenzoic acid, 97%, Thermo Scientific™

Catalog Number p-7031292
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1017778-60-7
C8H6F2O2
172.131
MFCD09832332
RNAYJQUWVKDTJZ-UHFFFAOYSA-N
3,4-difluoro-5-methylbenzoic acid, jrd
20112149
3,4-difluoro-5-methylbenzoic acid
CC1=C(C(=CC(=C1)C(=O)O)F)F
This item is not returnable. View return policy
1017778-60-7
C8H6F2O2
172.131
MFCD09832332
RNAYJQUWVKDTJZ-UHFFFAOYSA-N
3,4-difluoro-5-methylbenzoic acid, jrd
20112149
3,4-difluoro-5-methylbenzoic acid
CC1=C(C(=CC(=C1)C(=O)O)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 171°C to 175°C
Quantity 250 mg
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 3,4-Difluoro-5-methylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'