missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-2-methylbenzoic acid, 97+%, Alfa Aesar™

Brand:  Alfa Aesar H32326.06

Code : NEW

Additional Details : CAS Number : 157652-31-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15405047

  • £114.80 / 5g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Description

Description

Applications
3,4-Difluoro-2-methylbenzoic acid, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in medicine field, electronic industry and polymer material.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Specifications

Chemical Identifiers

3,4-Difluoro-2-methylbenzoic acid
C8H6F2O2
MFCD07777153
157652-31-8
172.13
3,4-Difluoro-o-toluic acid

Specifications

3,4-Difluoro-2-methylbenzoic acid
White to Cream
5g
Powder
C8H6F2O2
MFCD07777153
Odorless
152° to 156°C
157652-31-8
≥97%
172.13
3,4-Difluoro-o-toluic acid
Videos
SDS