missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylacetonitrile, 98%, ACROS Organics™

Brand:  Acros Organics 213941000

Code : 29

Additional Details : CAS Number : 3218-49-3 Weight : 0.15000kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10741871

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

3,4-Dichlorophenylacetonitrile
3218-49-3
100.0
98%
Cl2C6H3CH2CN
MFCD00001909
98%
97.5
98%
C8H5Cl2N
186.04
3, 4-Dichlorobenzyl cyanide

Specifications

3,4-Dichlorophenylacetonitrile
170.0°C (12.0 mmHg)
38.0° to 41.0°C
98%
97.5
Authentic
98%
C8H5Cl2N
186.04
Glass bottle
3, 4-Dichlorobenzyl cyanide
>110°C
White
100g
3218-49-3
100.0
Crystalline Powder
98%
Cl2C6H3CH2CN
MFCD00001909
09, III, 2272
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,4-Dichlorophenylacetonitrile

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS