missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7024486
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
6590-94-9
C7H3Cl2NS
204.07
MFCD00037828
OSBIEFWIIINTNJ-UHFFFAOYSA-N
3,4-dichlorophenyl isothiocyanate, benzene, 1,2-dichloro-4-isothiocyanato, 3,4-dichlorophenylisothiocyanate, benzene, 1,2-dichloro-4-isothiocyanato-9ci, isothiocyanic acid, 3,4-dichlorophenyl ester, 1,2-dichloro-4-isothiocyanato-benzene, acmc-20anfc, 4-12-00-01271 beilstein handbook reference, 3,4-dichloro-1-isothiocyanato-benzene, #
23065
1,2-dichloro-4-isothiocyanatobenzene
ClC1=CC=C(C=C1Cl)N=C=S
This item is not returnable. View return policy
6590-94-9
C7H3Cl2NS
204.07
MFCD00037828
OSBIEFWIIINTNJ-UHFFFAOYSA-N
3,4-dichlorophenyl isothiocyanate, benzene, 1,2-dichloro-4-isothiocyanato, 3,4-dichlorophenylisothiocyanate, benzene, 1,2-dichloro-4-isothiocyanato-9ci, isothiocyanic acid, 3,4-dichlorophenyl ester, 1,2-dichloro-4-isothiocyanato-benzene, acmc-20anfc, 4-12-00-01271 beilstein handbook reference, 3,4-dichloro-1-isothiocyanato-benzene, #
23065
1,2-dichloro-4-isothiocyanatobenzene
ClC1=CC=C(C=C1Cl)N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.422
Boiling Point 145°C to 148°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Refractive Index 1.68
Quantity 5 g
UN Number UN2922
Beilstein 511101
Sensitivity Moisture sensitive
Formula Weight 204.08
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'