missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenyl isocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7023170
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
102-36-3
C7H3Cl2NO
188.007
MFCD00002017
MFUVCHZWGSJKEQ-UHFFFAOYSA-N
3,4-dichlorophenyl isocyanate, benzene, 1,2-dichloro-4-isocyanato, 3,4-dichlorfenylisokyanat, isocyanic acid 3,4-dichlorophenyl ester, 3,4-dichlorophenylisocyanate, unii-ozw0b7f1du, 3,4-dichlorphenylisocyanate, isocyanic acid, 3,4-dichlorophenyl ester, 3,4-dichlorfenylisokyanat czech, ozw0b7f1du
7607
1,2-dichloro-4-isocyanatobenzene
C1=CC(=C(C=C1N=C=O)Cl)Cl
This item is not returnable. View return policy
102-36-3
C7H3Cl2NO
188.007
MFCD00002017
MFUVCHZWGSJKEQ-UHFFFAOYSA-N
3,4-dichlorophenyl isocyanate, benzene, 1,2-dichloro-4-isocyanato, 3,4-dichlorfenylisokyanat, isocyanic acid 3,4-dichlorophenyl ester, 3,4-dichlorophenylisocyanate, unii-ozw0b7f1du, 3,4-dichlorphenylisocyanate, isocyanic acid, 3,4-dichlorophenyl ester, 3,4-dichlorfenylisokyanat czech, ozw0b7f1du
7607
1,2-dichloro-4-isocyanatobenzene
C1=CC(=C(C=C1N=C=O)Cl)Cl

Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C 'labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C ′″labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, amines, alcohols.
TRUSTED_SUSTAINABILITY
Melting Point 40°C to 44°C
Boiling Point 118°C to 120°C (18 mmHg)
Flash Point 123°C (253°F)
Odor Irritating
Quantity 5 g
UN Number UN2250
Beilstein 608325
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 188.01
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorophenyl isocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'