missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenoxyacetic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-4228589
Click to view available options
Quantity:
5 g
10 g
This item is not returnable. View return policy
588-22-7
C8H6Cl2O3
221.03
MFCD00004303
SNYRXHULAWEECU-UHFFFAOYSA-N
3,4-dichlorophenoxyacetic acid, 2-3,4-dichlorophenoxy acetic acid, 3,4-dichlorophenoxy acetic acid, acetic acid, 3,4-dichlorophenoxy, acetic acid,4-dichlorophenoxy, acmc-20anfd, 4-06-00-00954 beilstein handbook reference, 3,4-dichloro-phenoxy-acetic acid, acetic acid,2-3,4-dichlorophenoxy
11495
CHEBI:393747
2-(3,4-dichlorophenoxy)acetic acid
OC(=O)COC1=CC=C(Cl)C(Cl)=C1
This item is not returnable. View return policy
588-22-7
C8H6Cl2O3
221.03
MFCD00004303
SNYRXHULAWEECU-UHFFFAOYSA-N
3,4-dichlorophenoxyacetic acid, 2-3,4-dichlorophenoxy acetic acid, 3,4-dichlorophenoxy acetic acid, acetic acid, 3,4-dichlorophenoxy, acetic acid,4-dichlorophenoxy, acmc-20anfd, 4-06-00-00954 beilstein handbook reference, 3,4-dichloro-phenoxy-acetic acid, acetic acid,2-3,4-dichlorophenoxy
11495
CHEBI:393747
2-(3,4-dichlorophenoxy)acetic acid
OC(=O)COC1=CC=C(Cl)C(Cl)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 138.0°C to 142.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Linear Formula Cl2C6H3OCH2CO2H
Quantity 5 g
Formula Weight 221.04
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dichlorophenoxyacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichlorophenoxyacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'