missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloronitrobenzene, 95%, Thermo Scientific™

Catalog Number 10736331
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
99-54-7
C6H3Cl2NO2
192.00
MFCD00007207
NTBYINQTYWZXLH-UHFFFAOYSA-N
3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech
7443
1,2-dichloro-4-nitrobenzene
[O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1
This item is not returnable. View return policy
99-54-7
C6H3Cl2NO2
192.00
MFCD00007207
NTBYINQTYWZXLH-UHFFFAOYSA-N
3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech
7443
1,2-dichloro-4-nitrobenzene
[O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1
CAS Min % 94.0
CAS Max % 100.0
Melting Point 39°C to 45°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula Cl2C6H3NO2
Quantity 1 kg
Beilstein 05, 246
Formula Weight 192
Percent Purity ≥94%
Physical Form Crystalline Mass
Chemical Name or Material 3, 4-Dichloronitrobenzene, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichloronitrobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'