missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloronitrobenzene, 95%, ACROS Organics™

£12.47 - £64.61

Chemical Identifiers

CAS 99-54-7
Molecular Formula C6H3Cl2NO2
Molecular Weight (g/mol) 192.00
MDL Number MFCD00007207
InChI Key NTBYINQTYWZXLH-UHFFFAOYSA-N
Synonym 3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech
PubChem CID 7443
IUPAC Name 1,2-dichloro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10010860
SDS View Documents
Acros Organics
113530050
5g Glass bottle
£12.47
5g
View alternative products
Save Up to 
»
Estimated Shipment: 10-08-2021
Please to purchase this item. Need a web account? Register with us today!
10666872
SDS View Documents
Acros Organics
113532500
250g Glass bottle
£23.06
250g
View alternative products
Save Up to 
»
Estimated Shipment: 10-08-2021
Please to purchase this item. Need a web account? Register with us today!
10736331
SDS View Documents
Acros Organics
113530010
1kg Glass bottle
£64.61
1kg
View alternative products
Save Up to 
»
Estimated Shipment: 10-08-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

99-54-7
192.00
NTBYINQTYWZXLH-UHFFFAOYSA-N
7443
[O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1
C6H3Cl2NO2
MFCD00007207
3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech
1,2-dichloro-4-nitrobenzene

Specifications

3, 4-Dichloronitrobenzene
Authentic
255°C to 256°C
39°C to 45°C
95%
94.0
94% min. (GC)
Cl2C6H3NO2
05, 246
Solubility in water: 151mg/l (20°C). Other solubilities: soluble in hot alcohol and ether
[O-][N+](=O)C1=CC=C(Cl)C(Cl)=C1
192.00
192
95%
123°C
Glass bottle
Yellow to Brown
5g
99-54-7
100.0
C6H3Cl2NO2
MFCD00007207
3,4-dichloronitrobenzene, benzene, 1,2-dichloro-4-nitro, dcnb, 1-nitro-3,4-dichlorobenzene, 3,4-dichlornitrobenzen, unii-ijt74ji7fs, ccris 3097, 1,2-dichloro-4-nitro-benzene, 3,4-dichlornitrobenzen czech, 3,4-dichloronitrobenzen czech
NTBYINQTYWZXLH-UHFFFAOYSA-N
1,2-dichloro-4-nitrobenzene
7443
Crystalline Mass
Safety and Handling

Safety and Handling

Product Identifier
  • 3,4-Dichloronitrobenzene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Product Certifications