missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorocinnamic acid, 97%, Thermo Scientific™

Catalog Number p-4258939
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1202-39-7
C9H6Cl2O2
217.05
RRLUFPHCTSFKNR-DUXPYHPUSA-N
3,4-dichlorocinnamic acid, e-3-3,4-dichlorophenyl acrylic acid, 3',4'-dichlorocinnamic acid, 2-propenoic acid, 3-3,4-dichlorophenyl, cinnamic acid, 3,4-dichloro, 3-3,4-dichlorophenyl acrylic acid, 2e-3-3,4-dichlorophenyl prop-2-enoic acid, 3-3,4-dichlorophenyl-2-propenoic acid, e-3-3,4-dichlorophenyl prop-2-enoic acid, 2e-3-3,4-dichlorophenyl acrylic acid
688027
(E)-3-(3,4-dichlorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1C=CC(=O)O)Cl)Cl
This item is not returnable. View return policy
1202-39-7
C9H6Cl2O2
217.05
RRLUFPHCTSFKNR-DUXPYHPUSA-N
3,4-dichlorocinnamic acid, e-3-3,4-dichlorophenyl acrylic acid, 3',4'-dichlorocinnamic acid, 2-propenoic acid, 3-3,4-dichlorophenyl, cinnamic acid, 3,4-dichloro, 3-3,4-dichlorophenyl acrylic acid, 2e-3-3,4-dichlorophenyl prop-2-enoic acid, 3-3,4-dichlorophenyl-2-propenoic acid, e-3-3,4-dichlorophenyl prop-2-enoic acid, 2e-3-3,4-dichlorophenyl acrylic acid
688027
(E)-3-(3,4-dichlorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1C=CC(=O)O)Cl)Cl
CAS Min % 96.0
CAS Max % 100.0
Melting Point 220.0°C to 223.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Linear Formula Cl2C6H3CH=CHCO2H
Quantity 5 g
Formula Weight 217.05
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 3, 4-Dichlorocinnamic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichlorocinnamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'