missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dichlorobiphenyl-4-sulfonyl chloride, 95%, Thermo Scientific™

Catalog Number 15224824
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
501697-80-9
C12H7Cl3O2S
321.61
DHVBZUPMGVOIOH-UHFFFAOYSA-N
3',4'-dichloro 1,1'-biphenyl-4-sulfonyl chloride, 4-3,4-dichlorophenyl benzenesulfonyl chloride, 3',4'-dichloro-biphenyl-4-sulfonyl chloride, 3',4'-dichloro-1,1'-biphenyl-4-sulfonyl chloride, 4-3,4-dichlorophenyl phenyl chlorosulfone, 4-3,4-dichlorophenyl phenyl sulfonyl chloride, 4-3,4-dichlorophenyl phenyl sulphonyl chloride
2794751
4-(3,4-dichlorophenyl)benzenesulfonyl chloride
C1=CC(=CC=C1C2=CC(=C(C=C2)Cl)Cl)S(=O)(=O)Cl
This item is not returnable. View return policy
501697-80-9
C12H7Cl3O2S
321.61
DHVBZUPMGVOIOH-UHFFFAOYSA-N
3',4'-dichloro 1,1'-biphenyl-4-sulfonyl chloride, 4-3,4-dichlorophenyl benzenesulfonyl chloride, 3',4'-dichloro-biphenyl-4-sulfonyl chloride, 3',4'-dichloro-1,1'-biphenyl-4-sulfonyl chloride, 4-3,4-dichlorophenyl phenyl chlorosulfone, 4-3,4-dichlorophenyl phenyl sulfonyl chloride, 4-3,4-dichlorophenyl phenyl sulphonyl chloride
2794751
4-(3,4-dichlorophenyl)benzenesulfonyl chloride
C1=CC(=CC=C1C2=CC(=C(C=C2)Cl)Cl)S(=O)(=O)Cl
Melting Point 104°C to 108°C
Assay Percent Range 94% min. Diethylsulfonamide derivative (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 321.61
Percent Purity 95%
Chemical Name or Material 3', 4'-Dichlorobiphenyl-4-sulfonyl chloride
compliance-icons
Product Identifier
  • 3',4'-Dichlorobiphenyl-4-sulfonyl chloride, 95%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'