missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl isothiocyanate, 97%, Thermo Scientific™

Catalog Number p-7058375
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
18967-42-5
C8H5Cl2NS
218.095
MFCD00041113
DPSSHXWTNIGVMQ-UHFFFAOYSA-N
3,4-dichlorobenzyl isothiocyanate, 1,2-dichloro-4-isothiocyanatomethyl benzene, isothiocyanic acid, 3,4-dichlorobenzyl ester, benzene, 1,2-dichloro-4-isothiocyanatomethyl, acmc-1bqjy, wln: scn1r cg dg, 4-12-00-02393 beilstein handbook reference, 3,4-dichlorobenzylisothiocyanate, 3,4-dichlorophenyl methanisothiocyanate, isothiocyanic acid,4-dichlorobenzyl ester
29350
1,2-dichloro-4-(isothiocyanatomethyl)benzene
C1=CC(=C(C=C1CN=C=S)Cl)Cl
This item is not returnable. View return policy
18967-42-5
C8H5Cl2NS
218.095
MFCD00041113
DPSSHXWTNIGVMQ-UHFFFAOYSA-N
3,4-dichlorobenzyl isothiocyanate, 1,2-dichloro-4-isothiocyanatomethyl benzene, isothiocyanic acid, 3,4-dichlorobenzyl ester, benzene, 1,2-dichloro-4-isothiocyanatomethyl, acmc-1bqjy, wln: scn1r cg dg, 4-12-00-02393 beilstein handbook reference, 3,4-dichlorobenzylisothiocyanate, 3,4-dichlorophenyl methanisothiocyanate, isothiocyanic acid,4-dichlorobenzyl ester
29350
1,2-dichloro-4-(isothiocyanatomethyl)benzene
C1=CC(=C(C=C1CN=C=S)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 26°C to 28°C
Density 1.38
Boiling Point 130°C (1 mmHg)
Odor Irritating
Refractive Index 1.6295
Quantity 1 g
UN Number UN2923
Beilstein 2937608
Sensitivity Moisture sensitive
Formula Weight 218.11
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzyl isothiocyanate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'