missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog Number p-3736167
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
18880-04-1
C7H5BrCl2
239.93
MFCD00041363
XLWSBDFQAJXCQX-UHFFFAOYSA-N
3,4-dichlorobenzyl bromide, 4-bromomethyl-1,2-dichlorobenzene, benzene, 4-bromomethyl-1,2-dichloro, 3,4-dichlorobenzylbromide, alpha-bromo-3,4-dichlorotoluene, acmc-209erq, 3,4 dichlorobenzyl bromide, 3,4-dichloro-benzyl bromide, ksc493a6b, timtec-bb sbb005763
639726
4-(bromomethyl)-1,2-dichlorobenzene
C1=CC(=C(C=C1CBr)Cl)Cl
This item is not returnable. View return policy
18880-04-1
C7H5BrCl2
239.93
MFCD00041363
XLWSBDFQAJXCQX-UHFFFAOYSA-N
3,4-dichlorobenzyl bromide, 4-bromomethyl-1,2-dichlorobenzene, benzene, 4-bromomethyl-1,2-dichloro, 3,4-dichlorobenzylbromide, alpha-bromo-3,4-dichlorotoluene, acmc-209erq, 3,4 dichlorobenzyl bromide, 3,4-dichloro-benzyl bromide, ksc493a6b, timtec-bb sbb005763
639726
4-(bromomethyl)-1,2-dichlorobenzene
C1=CC(=C(C=C1CBr)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.6500g/mL
Boiling Point 115°C to 120°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6050 to 1.607
Quantity 25 g
Specific Gravity 1.65
Formula Weight 239.93
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dichlorobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichlorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'