missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzoyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7023445
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
3024-72-4
C7H3Cl3O
209.45
MFCD00000672
VTXNOVCTHUBABW-UHFFFAOYSA-N
76403
3,4-dichlorobenzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)Cl)Cl
This item is not returnable. View return policy
3024-72-4
C7H3Cl3O
209.45
MFCD00000672
VTXNOVCTHUBABW-UHFFFAOYSA-N
76403
3,4-dichlorobenzoyl chloride
C1=CC(=C(C=C1C(=O)Cl)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 30°C to 34°C
Boiling Point 242°C
Flash Point 142°C (287°F)
Quantity 25 g
UN Number UN3261
Beilstein 607485
Synonym benzoyl chloride, 3,4-dichloro, 3,4-dichlorobenzoylchloride, 3,4-dichloro benzoyl chloride, 2,3-dichlorobenzoyl, 3,4dichlorobenzoyl chloride, acmc-1aj45, 3,4 dichlorobenzoyl chloride, 3,4-dichlorobenzoyl-chloride, ksc225o5d, 3,4-dichloro-benzoyl chloride
Formula Weight 209.46
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzoyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'