missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzoic acid, 98+%, ACROS Organics™

Catalog Number p-3523308
Click to view available options
Quantity:
25 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
51-44-5
C7H4Cl2O2
191.01
MFCD00002492
VPHHJAOJUJHJKD-UHFFFAOYSA-N
benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609
5817
CHEBI:49401
3,4-dichlorobenzoic acid
OC(=O)C1=CC=C(Cl)C(Cl)=C1
This item is not returnable. View return policy
51-44-5
C7H4Cl2O2
191.01
MFCD00002492
VPHHJAOJUJHJKD-UHFFFAOYSA-N
benzoic acid, 3,4-dichloro, unii-ys4zr9bxwx, 3,4-dichloro-benzoic acid, ys4zr9bxwx, benzoic acid,4-dichloro, wln: qvr cg dg, pubchem3686, acmc-209kts, dsstox_cid_4980, dsstox_rid_77609
5817
CHEBI:49401
3,4-dichlorobenzoic acid
OC(=O)C1=CC=C(Cl)C(Cl)=C1
CAS Min % 98.0
CAS Max % 100.0
Melting Point 205.0°C to 209.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Linear Formula Cl2C6H3CO2H
Quantity 25 g
Formula Weight 191.01
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 3, 4-Dichlorobenzoic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'