missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzenethiol, 97%, ACROS Organics™

Brand:  Acros Organics 162850050

Code : 29

Additional Details : CAS Number : 5858-17-3 Weight : 0.05500kg Transport : UN number : 2810 Chem class : 1 Pack group : III


Quantity 5g

 View more versions of this product

Product Code. 10881861

  • £42.40 / 5g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

3, 4-Dichlorobenzenethiol
96.0
97%
C6H4Cl2S
179.06
5858-17-3
100.0
97%
Cl2C6H3SH
MFCD00004840

Specifications

3, 4-Dichlorobenzenethiol
1.4
Cloudy colorless to yellow or white to yellow
5858-17-3
100.0
Authentic
Liquid or Low Melting Solid
97%
Cl2C6H3SH
MFCD00004840
>112°C
119.0° to 120.0°C (10.0 mmHg)
5g
96.0
1.4000g/mL
1.625
97%
C6H4Cl2S
179.06
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3,4-Dichlorothiophenol, 97%

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS