missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzenethiol, 97%, Thermo Scientific™

Catalog Number p-4228584
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
5858-17-3
C6H4Cl2S
179.06
MFCD00004840
HNJZDPKMMZXSKT-UHFFFAOYSA-N
3,4-dichlorothiophenol, 3,4-dichloro thiophenol, benzenethiol, 3,4-dichloro, 3,4-dichlorobenzene-1-thiol, pubchem6803, 3,4dichloro-thiophenol, acmc-1aset, 3,4-dichloro-thiophenol, 3 ,4-dichlorobenzenethiol, 3,4-dichlorophenylmercaptan
79964
3,4-dichlorobenzenethiol
C1=CC(=C(C=C1S)Cl)Cl
This item is not returnable. View return policy
5858-17-3
C6H4Cl2S
179.06
MFCD00004840
HNJZDPKMMZXSKT-UHFFFAOYSA-N
3,4-dichlorothiophenol, 3,4-dichloro thiophenol, benzenethiol, 3,4-dichloro, 3,4-dichlorobenzene-1-thiol, pubchem6803, 3,4dichloro-thiophenol, acmc-1aset, 3,4-dichloro-thiophenol, 3 ,4-dichlorobenzenethiol, 3,4-dichlorophenylmercaptan
79964
3,4-dichlorobenzenethiol
C1=CC(=C(C=C1S)Cl)Cl
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow or White to Yellow
Density 1.4000g/mL
Boiling Point 119.0°C to 120.0°C (10.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula Cl2C6H3SH
Refractive Index 1.625
Quantity 1 g
Specific Gravity 1.4
Formula Weight 179.06
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3, 4-Dichlorobenzenethiol
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dichlorothiophenol, 97%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'