missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4-Dichloro-D-phenylalanine, 97%

Brand:  Alfa Aesar™ H63217.06

Code : NEW

Additional Details : CAS Number : 52794-98-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15489788

  • £117.66 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3,4-Dichloro-D-phenylalanine
234.08
H-D-Phe(3,4-Cl2)-OH; (R)-2-Amino-3-(3,4-dichlorophenyl)propionic acid
52794-98-6
MFCD00671682

Specifications

3,4-Dichloro-D-phenylalanine
Solid
234.08
H-D-Phe(3,4-Cl2)-OH; (R)-2-Amino-3-(3,4-dichlorophenyl)propionic acid
52794-98-6
97%
MFCD00671682
5g
Videos
SDS