missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dibromothiophene, 97%, Thermo Scientific Chemicals

Catalog Number 10742101
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3141-26-2
C4H2Br2S
241.93
MFCD00005465
VGKLVWTVCUDISO-UHFFFAOYSA-N
18452
3,4-dibromothiophene
BrC1=CSC=C1Br
This item is not returnable. View return policy
3141-26-2
C4H2Br2S
241.93
MFCD00005465
VGKLVWTVCUDISO-UHFFFAOYSA-N
18452
3,4-dibromothiophene
BrC1=CSC=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 4°C to 5°C
Color Colorless to Yellow
Density 2.1300g/mL
Boiling Point 221°C to 222°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6390 to 1.641
Quantity 1 g
Specific Gravity 2.13
Synonym thiophene, 3,4-dibromo, 3,4-dibromo-thiophene, pubchem5510, thiophene,4-dibromo, 3, 4-dibromothiophene, acmc-1clwy, 3,4-bis bromanyl thiophene, ksc224a6j, 3,4-dibromothiophene, bidd:gt0493
Formula Weight 241.94
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 4-Dibromothiophene
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dibromothiophene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'