missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dibenzyloxybenzaldehyde, 99%, ACROS Organics™

£12.86 - £134.00

Chemical Identifiers

CAS 5447-02-9
Molecular Formula C21H18O3
Molecular Weight (g/mol) 318.36
MDL Number MFCD00004776
InChI Key XDDLXZHBWVFPRG-UHFFFAOYSA-N
Synonym 3,4-dibenzyloxybenzaldehyde, 3,4-bis benzyloxy benzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, xddlxzhbwvfprg-uhfffaoysa-n, 3.4-dibenzyloxybenzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa
PubChem CID 79526
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10518143
SDS View Documents
Acros Organics
112640010
1g Glass bottle
£12.86
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 17 Please to purchase this item. Need a web account? Register with us today!
10240803
SDS View Documents
Acros Organics
112640050
5g Glass bottle
£40.89
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 4 Please to purchase this item. Need a web account? Register with us today!
10005963
SDS View Documents
Acros Organics
112640250
25g Glass bottle
£134.00
25g
View alternative products
Save Up to 
»
Estimated Shipment
10-12-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

5447-02-9
318.36
XDDLXZHBWVFPRG-UHFFFAOYSA-N
79526
C21H18O3
MFCD00004776
3,4-dibenzyloxybenzaldehyde, 3,4-bis benzyloxy benzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, xddlxzhbwvfprg-uhfffaoysa-n, 3.4-dibenzyloxybenzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa

Specifications

3, 4-Dibenzyloxybenzaldehyde
Glass bottle
88°C to 92°C
5447-02-9
100.0
C21H18O3
MFCD00004776
XDDLXZHBWVFPRG-UHFFFAOYSA-N
79526
Powder
Authentic
White to Yellow
1g
98.5
98.5% min. (GC)
(C6H5CH2O)2C6H3CHO
3,4-dibenzyloxybenzaldehyde, 3,4-bis benzyloxy benzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, xddlxzhbwvfprg-uhfffaoysa-n, 3.4-dibenzyloxybenzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa
318.36
318.36
99%
SDS