missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Dibenzo[b,d]thienyl)benzeneboronic acid, 98%

Brand:  Alfa Aesar™ H64101.06

UNSPSC : 12141907

Code : NEW

Additional Details : CAS Number : 1307859-67-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15421149

  • £119.78 / 5g


Estimated Shipment date
from Supplier 10-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

3-(4-Dibenzo[b,d]thienyl)benzeneboronic acid
304.17
21494496
1307859-67-1
MFCD22200801
3-(4-Dibenzo[b,d]thienyl)phenylboronic acid

Specifications

3-(4-Dibenzo[b,d]thienyl)benzeneboronic acid
Solid
5g
MFCD22200801
3-(4-Dibenzo[b,d]thienyl)phenylboronic acid
1307859-67-1
98%
304.17
21494496
Documentation

Technical Documents